Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(Boc-amino)cyclohexanecarboxylic acid, 98%, Thermo Scientific™

Catalog No. AAH5940201
Change view
Click to view available options
Quantity:
0.25 g
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5940201 0.25 g
AAH59402ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5940201 Supplier Thermo Scientific Chemicals Supplier No. H5940201

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53292-89-0
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.30
MDL Number MFCD01862293,MFCD03453262,MFCD01862294
InChI Key KXMRDHPZQHAXML-UHFFFAOYSA-N
Synonym trans-4-boc-amino cyclohexanecarboxylic acid, cis-4-boc-amino cyclohexanecarboxylic acid, 4-boc-amino cyclohexanecarboxylic acid, cis-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, trans-4-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-4-aminocyclohexanecarboxylic acid, boc-1,4-trans-achc-oh, cis-4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 4-tert-butoxycarbonylamino cyclohexanecarboxylic acid
PubChem CID 2755996
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C(O)=O

Specifications

Color White
Beilstein 7641071
Quantity 0.25 g
Formula Weight 243.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material trans-4-(Boc-amino)cyclohexanecarboxylic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.