Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Bromostilbene 98.0+%, TCI America™
SDP

Catalog No. B24161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24161G 1 g
B24165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24161G Supplier TCI America Supplier No. B24161G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13041-70-8
Molecular Formula C14H11Br
Molecular Weight (g/mol) 259.146
MDL Number MFCD00155020
InChI Key ZZMMKLVIBZWGPK-VOTSOKGWSA-N
Synonym 4-bromostilbene, 1-bromo-4-styrylbenzene, e-1-bromo-4-styrylbenzene, 4-bromo-trans-stilbene, e-4-bromostilbene, 1-bromo-4-e-2-phenylethenyl benzene, z-4-bromostilbene, benzene, 1-bromo-4-1e-2-phenylethenyl, benzene, 1-bromo-4-2-phenylethenyl
PubChem CID 5374623
IUPAC Name 1-bromo-4-[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)Br

Specifications

Melting Point 140°C
Color White
Quantity 1 g
Formula Weight 259.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Bromostilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.