Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Hydroxy-L-proline 99.0+%, TCI America™
SDP

Catalog No. H029625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H029625G 25 g
H02965G 5 g
H0296100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H029625G Supplier TCI America Supplier No. H029625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-35-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00064320
InChI Key PMMYEEVYMWASQN-DMTCNVIQSA-N
Synonym trans-4-hydroxy-l-proline, hydroxyproline, l-hydroxyproline, 2s,4r-4-hydroxypyrrolidine-2-carboxylic acid, 4-hydroxy-l-proline, l-4-hydroxyproline, hydroxy-l-proline, trans-4-hydroxyproline, h-hyp-oh, hypro
PubChem CID 5810
ChEBI CHEBI:18095
IUPAC Name (2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 131.13
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Hydroxy-L-proline
EINECSNumber (9)-1628
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.