Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-Nitrostilbene 98.0+%, TCI America™
SDP

Catalog No. N054925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N054925G 25 g
N05495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N054925G Supplier TCI America Supplier No. N054925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1694-20-8
Molecular Formula C14H11NO2
Molecular Weight (g/mol) 225.247
MDL Number MFCD00053614
InChI Key ZISCOWXWCHUSMH-VOTSOKGWSA-N
PubChem CID 639659
ChEBI CHEBI:377106
IUPAC Name 1-nitro-4-[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 225.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Nitrostilbene
RTECSNumber WJ6950050
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.