Learn More
trans-Anethole, 98+%
Description
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-Anethole is used to inhibits lung and forestomach carcinogenesis, used as carbon and energy supplement in the culture media of Pseudomonas putida strain. It is also used as used as a flavoring substance.
Solubility
Soluble in benzene, ethyl acetate, acetone, carbon disulfide.
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 20°C to 23°C |
| Density | 0.991 |
| Boiling Point | 232°C to 235°C |
| Flash Point | 96°C (205°F) |
| Refractive Index | 1.56 |
| Quantity | 100 g |
| Beilstein | 774229 |
| Sensitivity | Light sensitive |
| Merck Index | 14,643 |
| Solubility Information | Soluble in benzene,ethyl acetate,acetone,carbon disulfide. |
| Show More |
Safety and Handling
| Hazard Category | H317 |
| Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
| Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
| EINECSNumber | 224-052-0 |
| RTECSNumber | BZ9275000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.