Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-beta-Styrylboronic acid pinacol ester, 98%

Catalog No. AAL1952903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1952903 1 g
AAL19529MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1952903 Supplier Thermo Scientific Chemicals Supplier No. L1952903
Add to Cart

CAS: 83947-56-2 | C14H19BO2 | 230.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83947-56-2
Molecular Formula C14H19BO2
Molecular Weight (g/mol) 230.11
MDL Number MFCD03453666
InChI Key ARAINKADEARZLZ-ZHACJKMWSA-N
Synonym e-4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, e-phenylethenylboronic acid, pinacol ester, trans-styrylboronic acid, pinacol ester, beta-styrylboronic acid pinacol ester, trans-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl styrene, 4,4,5,5-tetramethyl-2-styryl-1,3,2-dioxaborolane, trans-beta-styrylboronic acid pinacol ester, trans-2-phenylvinylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-e-2-phenylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-2-phenylvinyl-1,3,2-dioxaborolane
PubChem CID 5708413
SMILES CC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1

Specifications

Melting Point 27°C to 33°C
Boiling Point 128°C (4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Beilstein 8617179
IUPAC Name 4,4,5,5-tetramethyl-2-[(1E)-2-phenylethenyl]-1,3,2-dioxaborolane
Formula Weight 230.12
Percent Purity 99%
Chemical Name or Material trans-beta-Styrylboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.