Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-N,N'-Diacetylcyclohexane-1,2-diamine 98.0+%, TCI America™
SDP

Catalog No. D322625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D322625G 25 g
D32265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D322625G Supplier TCI America Supplier No. D322625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70924-78-6
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.266
InChI Key MZYLAHHTXWRMOL-NXEZZACHSA-N
Synonym trans-1,2-Bis(acetamido)cyclohexane
PubChem CID 11805689
IUPAC Name N-[(1R,2R)-2-acetamidocyclohexyl]acetamide
SMILES CC(=O)NC1CCCCC1NC(=O)C

Specifications

Melting Point 270°C
Color White-Yellow
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-N,N′-Diacetylcyclohexane-1,2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.