Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-o-Coumaric Acid 98.0+%, TCI America™
SDP

Catalog No. C039425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C039425G 25 g
C03945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C039425G Supplier TCI America Supplier No. C039425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 614-60-8
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004379
InChI Key PMOWTIHVNWZYFI-AATRIKPKSA-N
Synonym 2-hydroxycinnamic acid, o-coumaric acid, trans-2-hydroxycinnamic acid, 2-coumaric acid, trans-o-hydroxycinnamic acid, trans-o-coumaric acid, 2-coumarate, e-o-hydroxycinnamic acid, 2-hydroxycinnamate, o-hydroxy-trans-cinnamic acid
PubChem CID 637540
ChEBI CHEBI:18125
IUPAC Name (2E)-3-(2-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\C1=CC=CC=C1O

Specifications

Melting Point 217°C
Color White-Yellow
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-o-Coumaric Acid
RTECSNumber GD9090000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.