Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,4-Diphenyl-1,3-butadiene 99.0+%, TCI America™
SDP

Catalog No. D087925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D087925G 25 g
D08795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D087925G Supplier TCI America Supplier No. D087925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 886-65-7
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00004791
InChI Key JFLKFZNIIQFQBS-FNCQTZNRSA-N
Synonym 1e,3z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenylbutadiene, e,z-1,4-diphenyl-1,3-butadiene, cis,trans-1,4-diphenyl-1,3-butadiene, 1,3-butadiene, 1,4-diphenyl-, e,z, 1,4-diphenyl-1,3-butadiene, 1e,3z, benzene,1'-1,3-butadiene-1,4-diyl bis, 1z,3e-4-phenylbuta-1,3-dienyl benzene, benzene,1,1'-1,3-butadiene-1,4-diyl bis, 1,4-diphenyl-1,3-butadiene e,z-form mi
PubChem CID 1551381
IUPAC Name [(1E,3E)-4-phenylbuta-1,3-dien-1-yl]benzene
SMILES C(\C=C\C1=CC=CC=C1)=C/C1=CC=CC=C1

Specifications

Melting Point 152°C
Boiling Point 350°C
Quantity 25 g
Formula Weight 206.29
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,4-Diphenyl-1,3-butadiene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.