Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. D51271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51271G 1 g
D51275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51271G Supplier TCI America Supplier No. D51271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174350-08-4
Molecular Formula C21H30F2
Molecular Weight (g/mol) 320.468
InChI Key GDXLAJNVPUQWRF-UHFFFAOYSA-N
PubChem CID 22184030
IUPAC Name 1-[4-(4-ethylcyclohexyl)cyclohexyl]-2,3-difluoro-4-methylbenzene
SMILES CCC1CCC(CC1)C2CCC(CC2)C3=C(C(=C(C=C3)C)F)F

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 320.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4′-ethylbicyclohexyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.