Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl 98.0+%, TCI America™
SDP

Catalog No. M26385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26385G 5 g
M26381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26385G Supplier TCI America Supplier No. M26385G

Chemical Identifiers

CAS 155041-85-3
Molecular Formula C21H30
Molecular Weight (g/mol) 282.471
MDL Number MFCD12911573
InChI Key BCUMFFLWJNKNOU-UHFFFAOYSA-N
Synonym trans,trans-4-p-tolyl-4'-vinyl-1,1'-bi cyclohexane, trans,trans-4-p-tolyl-4'-vinyl-bicyclohexyl, 1-trans,trans-4'-ethenyl 1,1'-bicyclohexyl-4-yl-4-methylbenzene, trans,trans-4-4-methylphenyl-4'-vinylbicyclohexyl, 1-4-4-ethenylcyclohexyl cyclohexyl-4-methylbenzene, ccp-v 1, bcumfflwjnknou-ysxiemcbsa-n, 4-p-tolyl-4'-vinylbi cyclohexane, trans-4-p-tolyl-trans-4'-vinylbicyclohexyl, trans,trans-4-p-tolyl-4'-vinylbicyclohexyl
PubChem CID 20580010
IUPAC Name 1-[4-(4-ethenylcyclohexyl)cyclohexyl]-4-methylbenzene
SMILES CC1=CC=C(C=C1)C2CCC(CC2)C3CCC(CC3)C=C

Specifications

Melting Point 66°C
Color White
Quantity 5 g
Formula Weight 282.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(4-Methylphenyl)-4′-vinylbicyclohexyl
EINECSNumber (4)-1940
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.