Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam trans-Zeatin, Cytokinin Plant Growth Hormone, Solid, 50 mg

Catalog No. 502705087
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5087 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5087 Supplier Abcam Supplier No. AB14564050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Endogenous cytokinin plant growth hormone and potent AHK3 ligand.

  • Molecular Weight: 219.24 Da
  • Purity: >98%
  • Form: Solid
  • Class: Cytokinin (endogenous plant growth hormone)
  • Properties: Potent AHK3 ligand (Ki = 1.3 nM), senescence-delaying hormone, acts as a positional cue regulating lateral root spacing, mediates plant growth and development in vivo
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Substrate Properties Chemical Substrate
Target Molecule AHK3
Content And Storage Store at -20°C.
Shipping Condition Ambient
Buffer 100% trans-Zeatin
Form Solid
Formulation 100% trans-Zeatin
Quantity 50 mg
For Use With (Application) Plant growth and development research, lateral root spacing regulation, senescence-delaying, AHK3 ligand-mediated signaling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.