Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam trans-Zeatin Riboside, Cytokinin, >98% Purity, 100 mg
Description
- Purity: >98%
- Molecular Weight: 351.4 g/mol
- CAS Number: 6025-53-2
- Composition: 100% Trans-zeatin riboside
- Description: Naturally occurring cytokinin
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% trans-zeatin riboside |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Substrate Properties | Chemical Substrate |
| Formulation | 100% Trans-zeatin riboside |
| Quantity | 100 mg |
| For Use With (Application) | Plant biology, Cytokinin research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?