Learn More
TARGETMOL CHEMICALS INC TILDACERFONT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T262785MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tildacerfont is an antagonist of CRF1 and reduces the levels of adrenocorticotropic hormone and adrenal androgen. Tildacerfont can be used in studies about congenital adrenal hyperplasia. purity: 100%
Catalog No. 50-258-0711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.