Learn More
Market Lab TransCart Mobile Draw Cart Deluxe, ML7910
Supplier: Market Lab 7910
The TransCart Mobile Draw Cart Deluxe by MarketLab is a mobile medical cart with drawers and shelves for storing and transporting medical supplies and equipment. It has 5 drawers with full extension slides and dividers, 4 shelves, and 4 antimicrobial casters including 2 locking casters. The durable steel construction has a scratch resistant powder coated exterior. The cart has a weight capacity of 200 lbs. Overall dimensions are 41.5" H x 23.5" W x 22" D. This medical cart provides secure enclosed storage combined with easy mobility.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.