Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Transcription Termination Factor Mouse anti-Human, Mouse, Clone: 7B3H2, Proteintech
SDP

Catalog No. 501736336 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6336 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6336 Supplier Proteintech Group Inc Supplier No. 602351IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The antibody is specific to TTF1.

TTF1 is a transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin, thyroperoxidase, and thyrotropin receptor. Crucial in the maintece of the thyroid differentiation phenotype. May play a role in lung development and surfactant homeostasis.

Specifications

Antigen Transcription Termination Factor
Applications Western Blot
Classification Monoclonal
Clone 7B3H2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TTF1
Gene Accession No. Q15361, Q62187
Gene Alias TTF 1, TTF I, TTF1
Gene Symbols TTF1
Host Species Mouse
Immunogen Peptide
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22130, 7270
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.