Learn More
Promega Transfection Carrier DNA
Catalog No. PRE4881 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 x 20 μg
Catalog No. Quantity
PRE4881 5 x 20 μg
1 options
Catalog No. PRE4881 Supplier Promega Supplier No. E4881
A plasmid DNA used to reduce the amount of an expression vector or reporter vector in mammalian cell transfection without reducing the overall amount of DNA.
  • Plasmid DNA Used to Dilute Expression or Reporter Vectors for Transfection
  • Maintains the total amount of DNA used in transfection while reducing the amount of expression or reporter vector
  • Lowered expression of the protein of interest can overcome potential overexpression artifacts
  • Ratio of carrier DNA to reporter DNA may need to be optimized, depending on the application
  • Supplied at a concentration of 1μg/μL
Specifications
Quantity 5 x 20 μg
Content And Storage -30°C to -10°C