Learn More
Thermo Scientific™ Transfer Assembly Pump Tubing
GREENER_CHOICE
Catalog No. SH3134211
Change view
Click to view available options
Connector Types:
AseptiQuik G Connectors
Tubing Style:
1/2 in.TPE and Pumpsil Silicone
3/8 in. ID Silicone and Pumpsil
3/8 in. TPE and Pumpsil Silicone
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Tubing Style
SH3134211 AseptiQuik G Connectors 3/8 in. TPE and Pumpsil Silicone
SH3140611 AseptiQuik G Connectors 3/8 in. ID Silicone and Pumpsil
SH3134221 AseptiQuik G Connectors 1/2 in.TPE and Pumpsil Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. SH3134211 Supplier Thermo Scientific™ Supplier No. SH31342.11
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
TRUSTED_SUSTAINABILITY
Specifications
Description Transfer Assembly Pump Tubing, 3/8 inch ID TPE and Pumpsil Silicone, AseptiQuik G Connectors
Content And Storage Room Temperature
Quantity Each
Tubing Style 3/8 in. TPE and Pumpsil Silicone
Connector Types AseptiQuik G Connectors
Tubing Material Pharmaceutical-grade thermoplastic elastomer, platinum-cured silicone