Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Transfer Pipette Cup
SDP

Catalog No. 22000399
Change view
Click to view available options
Mounting:
Powerful Magnet Mount
Powerful VHB Tape Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-399 Powerful VHB Tape Mount
22-000-424 Powerful Magnet Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-399 Supplier Poltex Supplier No. TXFRCUPT
Add to Cart
Add to Cart

Store transfer pipettes up and out of the way.

Durable and cleanable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Transfer Pipettes, Swabs
Material PETG
Positions 1
Sterility Non-sterile
For Use With (Equipment) Transfer pipettes, swabs
Mounting Powerful VHB Tape Mount
Quantity 1
Length (Metric) 102 mm
Weight (Metric) 130 g
Height (Metric) 127 mm
Width (Metric) 84 mm
Show More Show Less

consumables not included

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.