Learn More
Eppendorf 3P Transfer Rack 1.5/2.0 mL
Supplier: Eppendorf 3P 3880000151
Transfer Rack 1.5/2.0 mL, for Eppendorf SmartBlock™ 1.5 mL and Eppendorf SmartBlock™ 2.0 mL. Holds up to 24 microtubes (1.5 mL or 2.0 mL) for easy transfer of multiple vessels in parallel. Convenient transfer of up to 24 hot vessels without burning the fingers. Temperature range from -86 °C to 121 °C for broad flexibility. Limited to ThermoMixer C and ThermoStat C.