Learn More
Eppendorf™ Transfer Rack for Eppendorf SmartBlock™
Catalog No. 05414272 Shop All Eppendorf North America Products
Change view
Click to view available options
Holds:
24 x 0.5 mL
24 x 1.5/2.0 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
05-414-272 24 x 1.5/2.0 mL
05-414-273 24 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 05-414-272 Supplier Eppendorf™ Supplier No. 3880000151
Add to Cart
Add to Cart
This item is not returnable. View return policy
Transfer rack suitable for individually sensor-controlled SmartBlock™ heating/cooling/mixing blocks.
  • Holds up to 24 microtubes for easy transfer of multiple vessels in parallel.
  • Convenient transfer of up to 24 hot vessels without burning the fingers
  • Temperature range from -86°C to 121°C for broad flexibility
  • Limited to ThermoMixer C annd ThermoStat C
TRUSTED_SUSTAINABILITY
Specifications
Holds 24 x 1.5/2.0 mL
For Use With (Equipment) SmartBlock, ThermoMixer
Temperature (Metric) Range −86°C to 121 °C