Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transferrin Antibody Pair [HRP]
SDP

Catalog No. NB009703 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009703 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009703 Supplier Novus Biologicals™ Supplier No. NBP27933215PLATES
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 15.63 pg/mL
Assay Sensitivity 15.63-1000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Transferrin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene ID (Entrez) 7018
Gene Symbol TF
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 45 ng of recombinant Human Transferrin.
  • Capture Antibody: 1 mg/mL of mouse anti-Human Transferrin monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human Transferrin monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target Transferrin
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.3 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.