Learn More
Athens Research And Technology Inc TRANSFERRIN APO 100MG
Supplier:  Athens Research And Technology Inc 1616A32001100MG
Encompass_Preferred
Apotransferrin, Human Plasma: 100 MG; Lyophilized from 10 mM phosphate buffer, pH 7.4.; MW 80,000; Purity: > =95% by SDS-PAGE; Iron Content: <30 ug per gram. Native; Purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-658
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.