Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transferrin Native Protein
SDP

Catalog No. NBP196856 Shop All R&D Systems Products
Change view
Click to view available options
:
1mg; Fluorescein
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP196856 1mg; Fluorescein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196856 Supplier Novus Biologicals™ Supplier No. NBP196856
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

Native Transferrin. Source: Serum The Transferrin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.

Specifications

Accession Number Q06AH7
For Use With (Application) Dot Blot, Immunocytochemistry/Immunofluorescence
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Gene ID (Entrez) 7018
Name Transferrin Protein
Storage Requirements 4°C in the dark.
Cross Reactivity Human
Purity or Quality Grade Multi-step
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.