Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free |
| Gene ID (Entrez) | 7018 |
| Name | Transferrin Protein |
| Quantity | 1 mg |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Rat |
| Product Line | Novus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
