Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Transferrin Native Protein
SDP

Catalog No. NBC104256 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC104256 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC104256 Supplier Novus Biologicals™ Supplier No. NBC104256
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Accession Number Q06AH7
For Use With (Application) Western Blot, ELISA, SDS-PAGE
Formulation Lyophilized from 0.2 M ammonium bicarbonate.
Gene ID (Entrez) 7018
Quantity 10 mg
Storage Requirements Store at 4°C.
Gene Alias Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Gene Symbol TF
Reconstitution Reconstitute with 1.0 mL deionized water.
Inhibitor Properties This product was lyophilized from 0.2 M ammonium bicarbonate for use in cell culture and assays. Transferrin has been tested in SDS-Page and is also suitable for use as antigen or ligand in immunochemical reactions, as a control or standard in assays, for conjugation and most other immunological methods requiring highly purified proteins.
Research Category Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Loading Controls, Stem Cell Markers
Purity or Quality Grade Multi-step
Protein Transferrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.