Learn More
Invitrogen™ Transferrin Receptor Monoclonal Antibody (MEM-75), Biotin
Shop All Thermo Scientific Products

Description
This antibody reacts with an extracellular epitope of CD71 antigen (transferrin receptor), a 95 kDa type II homodimeric transmembrane glycoprotein expressed on activated B and T lymphocytes, macrophages and erythroid precursors; it is lost on resting blood leukocytes. The antibody MEM-75 does not block binding of transferrin to the receptor.
Specifications
Specifications
| Antigen | Transferrin Receptor |
| Applications | Flow Cytometry, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MEM-75 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | TFRC |
| Gene Accession No. | P02786 |
| Gene Alias | 2610028K12Rik; AI195355; AI426448; AU015758; CD antigen CD71; CD71; chicken transferrin receptor; E430033M20Rik; I79_000642; IMD46; mammary tumor virus receptor 1; Mtvr1; Mtvr-1; p90; sTfR; T9; TfR; tfR1; TFR2; TFRC; TR; transferrin receptor; transferrin receptor (p90, CD71); transferrin receptor 2; transferrin receptor protein 1; Transferrin receptor protein 1, serum form; Trfr |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.