Learn More
Transgelin 2 Mouse anti-Human, Mouse, Clone: 2E5G1, Proteintech
Shop All Proteintech Group Inc Products

Description
Our test results showed that this antibody Is specific to TAGLN2. It does not bind TAGLN1 and TAGLN3.
Specifications
Specifications
| Antigen | Transgelin 2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 2E5G1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | TAGLN2 |
| Gene Accession No. | P37802, Q9WVA4 |
| Gene Alias | SM22 beta, SM22beta, TAGLN2, Transgelin 2, Transgelin 2/TAGLN2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.