Learn More
Diagnostic Biosystems TRANSGLUTAMINASE II Transglutaminase II
Supplier:  Diagnostic Biosystems MOB35305
Encompass_Preferred
This antibody reacts with a 77-85 kDa protein known as transglutaminase II (TGase II). TGase II catalyzes calcium-dependent post-translational modification of proteins by formation of an isopeptide bond within or between polypeptide chains. Different tissue and cell types express varying amounts of tissue transglutaminase.
Catalog No. 50-238-5343
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.