Learn More
XpressBio TRANSIL Membrane Affinity Kit, 96 Well Plate
SDP
Supplier:  XpressBio TMP04102096 96 WELL PL
Encompass_Preferred
The TRANSIL Membrane Affinity kit enables researchers to measure the interaction of drugs with a membrane. This in vitro assay is designed to measure the distribution coefficient of test compounds between phosphatidylcholine membranes and an aqueous buffer. We refer to this distribution coefficient as membrane affinity.
Catalog No. NC2333884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.