Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT™ Lentivirus System

Catalog No. MIR6655 Shop All Mirus Bio LLC Products
Change view
Click to view available options
No. of Reactions:
5
34
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MIR6655 34
MIR6650 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR6655 Supplier Mirus Bio™ Supplier No. MIR 6655
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TransIT™ Lentivirus System combines the benefits of the TransIT™-Lenti Transfection Reagent and the Lentivirus Packaging Mix Powered by MISSION™ Genomics with an optimized protocol to produce high functional lentivirus titers in HEK 293 cells.

  • High Performance - Provide higher functional titers
  • Simple Protocol - No media change required, single harvest
  • Animal Origin Free - Regulatory friendly

Specifications

Content And Storage -20°C
Cell Type 293-derived Cells
Format Kit
Sample Type Plasmid DNA
Promoter CMV
Transfection Technique Chemical Transfection
Concentration Proprietary
Vector pMISSION
Formulation Proprietary
For Use With (Application) Lentivirus production
No. of Reactions 34
Product Type Transfection reagent, lentivirus vectors, transduction reagent
Quantity 1 Ea.
Sterility Sterile
Volume (Metric) Sample Lenti Transfection Reagent: 1.5 mL, Lentivirus Packaging Mix: 34 x 1.7 mL, TransduceIT Reagent: 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.