Learn More
Mirus Bio TransIT-VirusGEN™ Transfection Reagent
Catalog No. MIR6720 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Transfection Reagent
Catalog No.
MIR6720 Transfection Reagent
1 options
This item is not returnable. View return policy
Catalog No. MIR6720 Supplier Mirus Bio Supplier No. MIR 6720
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
TransIT-VirusGEN™ Transfection Reagent is ideal for Recombinant Adeno-associated Virus and Lentivirus Production.
  • TransIT-VirusGEN Transfection Reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • Key benefits of TransIT-VirusGEN Transfection Reagent include:
  • Performance: Efficient DNA delivery for production of high-titer viral vectors
  • Scalability: Efficient production from < 1 ml to large scale cultures
  • Flexibility: Compatible with different recombinant virus and cell culture systems
  • Animal Origin Free: Fully synthetic transfection reagent formulation
  • TransIT-VirusGEN™ is ideal for academic research and biopharmaceutical discovery.
  • Titers can be further increased with optimized enhancers included in the VirusGEN AAV Transfection Kit and VirusGEN LV Transfection Kit.