Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ TransIT-VirusGEN™ Transfection Reagent

Catalog No. MIR6706 Shop All Mirus Bio LLC Products
Change view
Click to view available options
Quantity:
1 x 0.3 mL
1 x 0.75 mL
1 x 1.5 mL
5 x 1.5 mL
10 x 1.5 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MIR6706 10 x 1.5 mL
MIR6703 1 x 0.3 mL
MIR6704 1 x 0.75 mL
MIR6700 1 x 1.5 mL
MIR6705 5 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MIR6706 Supplier Mirus Bio™ Supplier No. MIR6706
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

TransIT-VirusGEN™ Transfection Reagent is ideal for recombinant adeno-associated virus and lentivirus production.

TransIT-VirusGEN™ Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.

  • Reliable – Consistent high virus titer production
  • Scalable – Efficient across different formats
  • Flexible – Compatible with different recombinant virus and cell culture systems
  • Animal Origin Free – High performance with maximum compatibility

Specifications

Content And Storage Store at -10°C to -30°C
Description Before each use, warm to room temperature and vortex gently.
Cell Type 293-derived Cells
Format Tube
Promoter None (Promoterless)
Transfection Technique Chemical Transfection
Product Type Transfection Reagent
Quantity 10 x 1.5 mL
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.