Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Transketolase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556165 Supplier Invitrogen™ Supplier No. PA556165
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HYYEGGIGEA VSSAVVGEPG ITVTHLAVNR VPRSGKPAEL LKMFGIDRDA IAQAVRGLIT KA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transketolase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TKT
Gene Accession No. P29401, P40142, P50137
Gene Alias epididymis luminal protein 107; epididymis secretory protein Li 48; HEL107; HEL-S-48; p68; TK; TKT; TKT1; Transketolase; transketolase (Wernicke-Korsakoff syndrome)
Gene Symbols TKT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TKT. Recombinant protein control fragment (Product #RP-92874).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21881, 64524, 7086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.