Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Transmissible Gastroenteritis Virus Monoclonal Antibody (TGEV10-116.3.3), Invitrogen™
Mouse Monoclonal Antibody
$450.00 - $1108.00
Specifications
Antigen | Transmissible Gastroenteritis Virus |
---|---|
Clone | TGEV10-116.3.3 |
Concentration | 1.66 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA546180
|
Invitrogen™
MA546180 |
100 μg |
Each for $450.00
|
|
|||||
01-671-813
|
Invitrogen™
MA110783 |
500 μg |
Each for $600.50
|
|
|||||
PIMA546145
|
Invitrogen™
MA546145 |
1 mg |
Each for $1,108.00
|
|
|||||
Description
MA1-10783 detects Transmissible Gastroenteritis Virus. MA1-10783 has been successfully used in ELISA applications. The MA1-10783 immunogen is TGEV-infected ST cells.
Transmissible gastroenteritis virus or Transmissible gastroenteritis coronavirus is a coronavirus which infects pigs. It is an enveloped, positive-sense, single-stranded RNA virus which enters its host cell by binding to the APN receptor.Specifications
Transmissible Gastroenteritis Virus | |
1.66 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
TGEV-infected ST cells. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
TGEV10-116.3.3 | |
ELISA | |
Unconjugated | |
Mouse | |
Virus | |
TGV | |
IgG1 | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title