Learn More
Hunter Lab 10mm Transmission Cell
Supplier: Hunter Lab 13857340
Optically clear glass cells with fixed path length of 10 mm, 20 mmor 50 mm. Sample volume 20 ml, 40 ml and 100 ml respectively.For use with Transmission Spill Tray and Cell Holder.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.