Learn More
Grainger Transport Drum, Polyethylene
Supplier:  Grainger 1MLC6
Encompass_Preferred
The Transport Drum Polyethylene by Grainger is a 55 gallon open head drum made of polyethylene plastic. It has a 2 inch NPS threaded bung closure and is UN rated for hazardous materials transport. The dimensions are 23" diameter x 35" height. It has a weight capacity of 440 lbs and a volume of 55 gallons. The color is natural polyethylene. This drum is ideal for storing and transporting chemicals, petroleum products, food ingredients and other liquids. It meets UN packaging standards.
Catalog No. NC2510540
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.