Learn More
Market Lab MarketLab Specimen Transport Tote, Green, Intermediate, 14""""W x 8.5""""D x 7""""H
Supplier:  Market Lab 8250
Encompass_Preferred
Insulated heavy-duty, cleanable PVC-coated 210D nylon. Liner wipes clean with 10% bleach solution. Safely accommodates ice packs or dry ice. Lockable zippers. Interior and exterior document pouches and windows.
Catalog No. NC1288098
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.