Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Transthyretin Recombinant Rabbit Monoclonal Antibody (24H23L8)

Catalog No. 703278
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-327-8 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-327-8 Supplier Invitrogen™ Supplier No. 703278
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine.

Transthyretin (TTR), generally called prealbumin, is a plasma protein that plays an important role in physiology such as a transporter of hormone thyroxine and retinal-binding protein. After produced primarily in the liver, TTR is excreted into the plasma. TTR represents a disproportionate fraction (25%) of CSF protein, prompting the suggestion that it is either selectively transported across the blood-CSF barrier or synthesized de novo within the central nervous system. Transthyretin is a constituent found to the neuritic plaques, neurofibrillary tangles, and microangiopathic lesions of senile cerebral amyloid. It has been reported that more than 40 different mutations in the TTR gene associated with amyloid deposition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Transthyretin
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 24H23L8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene TTR
Gene Accession No. P02766
Gene Alias AA408768; AI787086; Amyloidosis I; ATTR; carpal tunnel syndrome 1; CTS; CTS1; D17860; epididymis luminal protein 111; HEL111; HsT2651; Lr1; PALB; prealbumin; pre-albumin; prealbumin, amyloidosis type I; Tbpa; thyroxine-binding prealbumin; transthyretin; Tt; TTR
Gene Symbols TTR
Host Species Rabbit
Immunogen Protein corresponding to human Transthyretin [aa21-aa147].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7276
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.