Learn More
CPC Scientific H-Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PARP023A
Encompass
This peptide can inhibit monocyte spreading on fibronectin-coated slides. The rapidly induced withdrawal of pseudopodial processes and rounding of the cells is accompanied by nitric oxide and endothelin-1 release. Spreading might be inhibited by autocrine endothelin-1 release and subsequent endothelin B receptor-dependent nitric oxide productionONE-LETTER SEQUENCE: SFLLRNPNDKYEPF-NH2MOLECULAR FORMULA: C81H119N21O22MOLECULAR WEIGHT:1738.96STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [141923-36-6], RESEARCH AREA: HematologyREFERENCES: K.D.Srivastava and H.I.Magazine, J. Immunol., 161, 5039 (1998); J.R. Ngaiza and E.A. Jaffe, Biochem. Biophys. Res. Commun., 179, 1656 (1991); D.M. Feng, D.F. Veber, T.M. Connolly, C. Condra, M.J. Tang, and R.F. Nutt, J. Med. Chem.,38,4125 (1995)
Catalog No. 50-211-0135
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.