Learn More
Medchemexpress LLC HY-P0078 10mg Medchemexpress, TRAP-6 CAS:141136-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP007810MG
Encompass_Preferred
Medchemexpress, HY-P0078 10mg TRAP-6 CAS:141136-83-6 TRAP-6 (PAR-1 agonist peptide), a peptide fragment, is a selective protease activating receptor 1 (PAR1) agonist. TRAP-6 activates human platelets via the thrombin receptor. TRAP-6 shows no activity at PAR4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3017
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.