Learn More
CPC Scientific H-Ser-Phe-Leu-Leu-Arg-Asn-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PARP019A
Encompass
This is thrombin receptor agonist peptide.SEQUENCE: H-Ser-Phe-Leu-Leu-Arg-Asn-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: SFLLRN-NH2MOLECULAR FORMULA: C34H57N11O8MOLECULAR WEIGHT: 747.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [141923-40-2]SYNONYMS: Thrombin Receptor Activator Peptide 6 amide, Thrombin Receptor (1-6) amide (human), PAR-1 (1-6) amide (human), Proteinase Activated Receptor 1 (1-6) amide (human), SFLLRN-NH2 Selective Protease-Activated Receptor 1 (PAR1) Agonist, SFLLRNamide, Coagulation Factor II Receptor (1-6) amide (human)RESEARCH AREA: Hematology, CardiovascularREFERENCES:R.M.Scarborough et al., J. Biol. Chem., 267, 13146 (1992)Cocks, T. et al. Gastroenterol. 116, 586 (1999)Mule, F. et al. J. Pharmacol. Exp. Ther. 303 1265 (2002)Koetzner, L. et al. J. Pharmacol. Exp. Ther. 10, 1124 (2004)Offermanns S. et al. Biochem. J. 290, 27 (1993)Selnick, H. et al. Curr. Med. Chem.- Cardio. Hematological Agents 1, 47 (2003)SKU(s): PARP-019A,
Catalog No. 50-211-0127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.