Learn More
CPC Scientific H-Ser-Phe-Leu-Leu-Arg-Asn-Pro-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific PARP021A
Encompass
This peptide is the fragment 42-48 of the thrombin receptor. TRAP-7 is as effective as thrombin in inducing Ca2+ mobilization and protein kinase C activation in some T cell lines.ONE-LETTER SEQUENCE: SFLLRNPMOLECULAR FORMULA: C39H63N11O10MOLECULAR WEIGHT:846.01STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [145229-76-1], RESEARCH AREA: HematologyREFERENCES: B.Mari et al., J. Biol. Chem., 269, 8517 (1994); Molloy, C. et al. J. Clin. Invest. 97, 1173 (1996); Ishida, A. et al. FEBS Lett. 427, 155 (1998); Ishida, A. and H. Fujisawa, J. Biol. Chem. 270, 2163 (1995)
Catalog No. 50-211-0131
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.