Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRAP alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552811 Supplier Invitrogen™ Supplier No. PA552811
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESRKRKRPIQ KVEMGTSSQN DVDMSWIPQE TLNQINKASP RRLPRKRAQK RSVGSDE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAP alpha
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SSR1
Gene Accession No. P43307, Q7TPJ0, Q9CY50
Gene Alias 2510001K09Rik; 6330400D04; Ac2-238; AI159733; AI452176; cb758; DKFZp781N23103; liver regeneration-related protein LRRG137; PGP35; PSEC0262; signal sequence receptor subunit 1; signal sequence receptor subunit alpha; signal sequence receptor, alpha; signal sequence receptor, alpha (translocon-associated protein alpha); SSR; SSR alpha subunit; SSR1; SSRA precursor protein; SSR-alpha; translocon associated protein alpha; translocon-associated protein alpha; translocon-associated protein alpha subunit; translocon-associated protein subunit alpha; TRAP alpha; TRAPA; TRAP-alpha; unnamed protein product
Gene Symbols SSR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSR1. Recombinant protein control fragment (Product #RP-90181).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107513, 361233, 6745
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.