Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAP1 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
SDP

Catalog No. 502540453
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0453 30 μL
50-167-4938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0453 Supplier OriGene Supplier No. TA504202S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRAP1 Monoclonal antibody specifically detects TRAP1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen TRAP1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ACD
Gene Accession No. Q96AP0
Gene Alias 2410002K23Rik; heat shock protein 75 kDa, mitochondrial; HSP 75; HSP75; HSP90L; testicular tissue protein Li 209; TNF receptor associated protein 1; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP1; TRAP-1; tumor necrosis factor type 1 receptor associated protein; tumor necrosis factor type 1 receptor-associated protein
Host Species Mouse
Immunogen Full length human recombinant protein of human ACD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65057
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.