Learn More
Invitrogen™ TRAPPC5 Monoclonal Antibody (3D9)

Description
Peptide Sequence: MEARFTRGKS ALLERALARP RTEVSLSAFA LLFSELVQHC QSRVFSVAEL QSRLAALGRQ VGARVLDALV AREKGARRET KVLGALLFVK GAVWKALFGK EADKLEQAND DARTFYIIER EPLINTYISV PKENSTLNCA SFTAGIVEAV LTHSGFPAKV TAHWHKGTTL MIKFEEAVIA RDRALEGR.
Specifications
Specifications
| Antigen | TRAPPC5 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3D9 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TRAPPC5 |
| Gene Accession No. | Q8IUR0 |
| Gene Alias | 4021401A16Rik; RGD1563971; trafficking protein particle complex 5; trafficking protein particle complex subunit 5; Trappc5; TRS31 |
| Gene Symbols | TRAPPC5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.