Learn More
TARGETMOL CHEMICALS INC TRASTUZUMAB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T9912
Encompass_Preferred
Also available in 1 mg 10 mg and bulk. Please contact Fisher for quotes. Trastuzumab is a humanized monoclonal antibody for patients with invasive breast cancers that overexpress HER2. Trastuzumab has been clinically used to treat HER2 Positive Metastatic Breast Cancer and HER2 Positive Gastric Cancer. purity: 99%
Catalog No. 50-257-8931
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.