Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trastuzumab Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9547U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9547U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9547U100 Supplier R&D Systems Supplier No. FAB95471U100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Trastuzumab Monoclonal specifically detects Trastuzumab in Multi-Species samples. It is validated for Flow Cytometry.

Specifications

Antigen Trastuzumab
Applications Flow Cytometry
Classification Monoclonal
Clone 2261E
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide
Host Species Rabbit
Immunogen Full-length Trastuzumab
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.