Learn More
Julabo USA Inc Each, tray and 4 test tube racks, 360 tubes 13mm Diameter. SW22/SW23.
SDP
Supplier:  Julabo USA Inc 8960441
Encompass_Preferred
Bath Accessories, Julabo, Tray with 4 racks 8970381 for 360 tubes 75 x 12/13 mm, to +80°C. For use with shaking water baths: SW22 / SW23.
Catalog No. 50-753-4072
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.