Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRBC2 Chimeric Recombinant Mouse Monoclonal Antibody (SAM.2), Alexa Fluor™ 647

Catalog No. 704905AF647
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-490-5AF647 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-490-5AF647 Supplier Invitrogen™ Supplier No. 704905AF647
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Recombinant Monoclonal Antibody

This is a recombinant antibody conjugated to Alexa Fluor™ 647 dye using SiteClick™ Antibody Labeling Technology. The SiteClick sDIBO Alkynes for Antibody Labeling employs copper-free click chemistry to covalently link a dibenzocyclooctyne (sDIBO) labeled Alexa Fluor™ 647 dye to an azido modified antibody. SiteClick™ technology allows site-specific conjugation of an antibody with complete confidence that the label, when attached to the antibody, is located far away from its antigen binding domain and will not interfere with the antibody function. Recombinant human-mouse chimeric monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

TRBC2 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRBC2 Chimeric
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone SAM.2
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 647
Formulation TBS with 0.02% sodium azide; pH 7.0
Gene TRBC2
Gene Accession No. A0A5B9
Gene Alias T-cell receptor beta-2 chain C region; TCRBC2; TRBC2
Gene Symbols TRBC2
Host Species Mouse
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28638
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.